Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Innovating Science™ Arabidopsis Seeds
SCI_ED

Catalog No. s37737
Click to view available options
Quantity:
300/Pk.

Ideal for the study the process of artificial selection. Innovating Science™ Arabidopsis Seeds are a fast Growing Plant Seeds

  • 300/Pack.
  • Light brown

Specifications

For Use With (Application) Study the process of artificial selection
Storage Requirements Room Temperature
Quantity 300/Pk.
Color Light Brown
Sufficient For Class of 60 students
Product Type Seed
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.