Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Arabidopsis thaliana ACS8 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5117718
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5117718 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5117718 Supplier Invitrogen™ Supplier No. PA5117718
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This antibody has been tested in direct-ELISA.

1-aminocyclopropane-1-carboxylate synthase (ACS) enzymes catalyze the conversion of S-adenosyl-L-methionine (SAM) into 1-aminocyclopropane-1-carboxylate (ACC), a direct precursor of ethylene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Arabidopsis thaliana ACS8
Applications Western Blot
Classification Polyclonal
Concentration 4.28 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene ACS8
Gene Accession No. Q9T065
Gene Alias 1-aminocyclopropane-1-carboxylate synthase - like protein; 1-amino-cyclopropane-1-carboxylate synthase 8; 1-aminocyclopropane-1-carboxylate synthase 8; 1-aminocyclopropane-1-carboxylate synthase ACS8; ACC synthase 8; ACS8; At4g37770; S-adenosyl-L-methionine methylthioadenosine-lyase 8; T28I19.50; T28I19_50
Gene Symbols ACS8
Host Species Rabbit
Immunogen Recombinant Arabidopsis thaliana 1-aminocyclopropane-1-carboxylate synthase 8 protein (1-469aa).
Purification Method Protein G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 829933
Target Species Plant
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.