Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Arabidopsis thaliana MYB75 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5117712
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5117712 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5117712 Supplier Invitrogen™ Supplier No. PA5117712
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This antibody has been tested in direct-ELISA.

Transcription activator, when associated with BHLH12/MYC1, EGL3, or GL3. Promotes the synthesis of. phenylpropanoid-derived compounds such as anthocyanins and proanthocyanidin, probably together with GL3 and BHLH2. Regulates the expression of CHS, DFRA, LDOX, and BAN.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Arabidopsis thaliana MYB75
Applications Western Blot
Classification Polyclonal
Concentration 3.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene MYB75
Gene Accession No. Q9FE25
Gene Alias At1g56650; ATMYB75; F25P12.92; F25P12_92; MYB DOMAIN PROTEIN 75; MYB75; Myb-related protein 75; MYELOBLASTOSIS PROTEIN 75; PAP1; phosphatidic acid phosphatase 1; production of anthocyanin pigment 1; Production of anthocyanin pigment 1 protein; SIAA1; SUC-INDUCED ANTHOCYANIN ACCUMULATION 1; Suc-induced anthocyanin accumulation locus 1; transcription factor; Transcription factor MYB75
Gene Symbols MYB75
Host Species Rabbit
Immunogen Recombinant Arabidopsis thaliana Transcription factor MYB75 protein (91-167aa).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 842120
Target Species Plant
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.