Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Arabidopsis thaliana OSM34 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5117589
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5117589 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5117589 Supplier Invitrogen™ Supplier No. PA5117589
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This antibody has been tested in direct-ELISA.

defense response to bacterium, incompatible interaction, defense response to fungus, incompatible interaction, response to salt stress.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Arabidopsis thaliana OSM34
Applications Western Blot
Classification Polyclonal
Concentration 3 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene OSM34
Gene Accession No. P50700
Gene Alias AT4G11650; ATOSM34; OSM34; osmotin 34; Osmotin-like protein OSM34; T5C23.80; T5C23_80
Gene Symbols OSM34
Host Species Rabbit
Immunogen Recombinant Arabidopsis thaliana Osmotin-like protein OSM34 protein (23-244aa).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 826770
Target Species Plant
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.