Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Arabidopsis thaliana PAP1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5117648
Description
This antibody has been tested in direct-ELISA.
Probably involved in light/cold stress-related jasmonate (JA) biosynthesis. Contributes to the protection of photosystem II (PSII) against light stress.
Specifications
Arabidopsis thaliana PAP1 | |
Polyclonal | |
Unconjugated | |
PAP1 | |
At4g04020; AtPap1; AtPGL35; FBN1a; FIB; FIB1a; fibrillin; fibrillin 1a; Fibrillin-1a; PAP1; PGL35; plastoglobulin 35; Probable plastid-lipid-associated protein 1, chloroplastic; putative fibrillin; T24H24.16; T24H24_16 | |
Rabbit | |
Protein G | |
RUO | |
825714 | |
-20°C or -80°C if preferred | |
Liquid |
Western Blot | |
4.221 mg/mL | |
PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
O81439 | |
PAP1 | |
Recombinant Arabidopsis thaliana Probable plastid-lipid-associated protein 1, chloroplastic protein (56-318aa). | |
100 μL | |
Primary | |
Plant | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction