Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Arabidopsis thaliana PMRT15 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5117690
Description
This antibody has been tested in direct-ELISA.
Methylates arginine residues of myelin basic protein (MBP) in vitro. Methylates symmetrically histone H4 of the FLC chromatin to form H4R3me2s, which in turn suppresses FLC expression to induce flowering. Regulates alternative splicing by methylating spliceosomal proteins. Involved in the post-transcriptional regulation of the circadian clock.
Specifications
| Arabidopsis thaliana PMRT15 | |
| Polyclonal | |
| Unconjugated | |
| PMRT15 | |
| At4g31120; AtPMRT15; ATPRMT5; calcium underaccumulation 1; CAU1; F6E21.40; F6E21_40; PMRT15; PMRT5; PRMT5; PROTEIN ARGININE METHYLTRANSFERASE 5; Protein arginine N-methyltransferase 1.5; SHK1 binding protein 1; Shk1 kinase-binding protein 1 homolog; SKB1 | |
| Rabbit | |
| Protein G | |
| RUO | |
| 829240 | |
| -20°C or -80°C if preferred | |
| Liquid |
| Western Blot | |
| 3.155 mg/mL | |
| PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
| Q8GWT4 | |
| PMRT15 | |
| Recombinant Arabidopsis thaliana Protein arginine N-methyltransferase 1.5 protein (513-637aa). | |
| 100 μg | |
| Primary | |
| Plant | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction