Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Arachidonic acid, ≥98%, MP Biomedicals™

Catalog No. MP215038490 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
500 mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP215038490 500 mg
ICN15038410 10 mg
MP215038450 50 mg
MP215038480 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. MP215038490 Supplier MP Biomedicals, Inc Supplier No. 0215038490
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Arachidonic Acid, >98%, Clear, Colorless Liquid, essential fatty acid, and a precursor in the biosynthesis of prostaglandins, thromboxanes, and leukotrienes.

  • Occurs in liver, brain, glandular organs, and depot fats of animals, in small amounts in human depot fats, and is a constituent of animal phosphatides.
  • Arachidonic acid (AA) is an unsaturated ω6 fatty acid constituent of the phospholipids of cell membranes.
  • Phospholipase A2 releases AA from the membrane phospholipids in response to inflammation.
  • AA is subsequently metabolized to prostaglandins and thromboxanes by at least two cyclooxygenase (COX) isoforms, to leukotrienes and lipoxins by lipoxygenases, and to epoxyeicosatrienoic acids via cytochrome p450-catalyzed metabolism.
  • AA and its metabolites play important roles in a variety of biological processes, including signal transduction, smooth muscle contraction, chemotaxis, cell proliferation and differentiation, and apoptosis.
  • AA has been demonstrated to bind to the a subunit of G protein and inhibit the activity of Ras GTPase-activating proteins (GAPs).
  • Cellular uptake of AA is energy dependent and involves protein-facilitated transport across the plasma membrane.
  • Arachidonic acid stimulates adhesion of MDA-MB-435 human metastatic cancer cells to extracellular matrix molecules (collagen IV and vitronectin).
  • Arachidonic acid plays a key role in cellular regulation and is controlled through multiple interconnected pathways.
  • Changes in arachidonic acid levels correspond to the synthesis of eicosanoids resulting from receptor-stimulated lipid hydrolysis.
  • It activates protein kinase C.
  • Arachidonic acid serves as a precursor to numerous eicosanoids as well as other bioactive molecules.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 506-32-1
Molecular Formula C20H32O2
MDL Number MFCD00004417

Specifications

Melting Point -49°C (literature)
Density 0.922 g/mL (Lit.)
Boiling Point 170°C at 0.2 mm Hg (Literature)
Refractive Index n20/D 1.4872(Lit.)
Quantity 500 mg
Synonym 5,8,11,14-Eicosatetraenoic Acid, (all-Z)-5,8,11,14-Eicosatetraenoic acid, cis,cis,cis,cis-5,8,11,14-Eicosatetraenoic acid, Eicosa-5Z,8Z,11Z,14Z-tetraenoic acid
Solubility Information If ethanol is undesirable, arachidonic acid may be dissolved in acetonitrile, DMF, or DMSO. Simply evaporate the ethanol under agentle stream of nitrogen (be certain not to evaporate the material to dryness) and redissolve the arachidonic acid in the solvent of choice. It is difficult to obtain aqueous solutions of arachidonic acid directly. However, an organic solvent free solution of arachidonic acid can be prepared using concentrated basic buffers (pH > 8.0 and ionic strength not less than 0.1 M). Add 400 µL of cold buffer (0°C) permg of arachidonic acid and agitate vigorously and/or ultrasonicate.
InChI Key YZXBAPSDXZZRGB-CGRWFSSPSA-N
Molecular Weight (g/mol) 304.47
Physical Form Liquid
Grade >98%
Show More Show Less
RTECSNumber CE6675000
storageNote1 -20°C, desiccate, protect from light, store under nitrogen
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.