Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ARAP1 Antibody, Alexa Fluor™ 594, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB10068223AF594
Description
ARAP1 Polyclonal antibody specifically detects ARAP1 in Human, Mouse samples. It is validated for Western BlotSpecifications
ARAP1 | |
Polyclonal | |
Western Blot | |
arf-GAP with Rho-GAP domain, ANK repeat and PH domain-containing protein 1, ArfGAP with RhoGAP domain, ankyrin repeat and PH domain 1, centaurin, delta 2, Centaurin-delta-2, CENTD2ARF-GAP, RHO-GAP, ankyrin repeat, and pleckstrin homology domains-containingprotein 1, cnt-d2, KIAA0782centaurin-delta-2 | |
The immunogen recognized by this antibody maps to a region between residue 1160 and 1210 of human centaurin, delta 2 (ankyrin repeat, and pleckstrin homology domains-containing protein 1) using the numbering given in entry NP_056057.1 (GeneID 116985). | |
0.1 mL | |
Cell Cycle and Replication | |
116985 | |
Store at 4C in the dark. | |
IgG |
Western Blot | |
Alexa Fluor 594 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human, Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction