Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ARAP1 Antibody (ARAP1-2), Alexa Fluor™ 532, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB11068801AF532
Description
ARAP1 Monoclonal specifically detects ARAP1 in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
ARAP1 | |
Monoclonal | |
Alexa Fluor 532 | |
50 mM sodium borate with 0.05% sodium azide | |
ARAP1 | |
His6-tagged protein corresponding to amino acids 1190-1450 of human ARAP1 | |
0.1 mL | |
Primary | |
The ARAP1-2 reacts with C-terminal part of ARAP1, a 160 kDa adaptor protein. | |
Store at 4°C in the dark. |
Western Blot, Immunocytochemistry, Immunofluorescence | |
ARAP1-2 | |
Western Blot, Immunocytochemistry/Immunofluorescence | |
arf-GAP with Rho-GAP domain, ANK repeat and PH domain-containing protein 1, ArfGAP with RhoGAP domain, ankyrin repeat and PH domain 1, centaurin, delta 2, Centaurin-delta-2, CENTD2ARF-GAP, RHO-GAP, ankyrin repeat, and pleckstrin homology domains-containingprotein 1, cnt-d2, KIAA0782centaurin-delta-2 | |
Mouse | |
Protein A purified | |
Cell Cycle and Replication | |
116985.0 | |
Human | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction