Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ARAP1 Monoclonal Antibody (ARAP1-2)
GREENER_CHOICE

Catalog No. MA119728
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA119728 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MA119728 Supplier Invitrogen™ Supplier No. MA119728
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This antibody reacts with C-terminal part of ARAP1 (intracellular epitope), a 160 kDa adaptor protein.

ARAP1 (angiotensin II type 1 receptor-associated protein) is an adaptor protein with ArfGAP and RhoGAP activities, containing five PH domains and ankyrin repeat. This adaptor seems to serve as a link between phosphoinositide- Arf-, and Rho-mediated cell signaling. ARAP1 supports the plasma membrane recycling of angiotensin II type 1 receptor (AT1) and is important also for cell-specific trafficking of pro-death receptor TRAIL-R1 (DR4) to the plasma membrane, thus promoting the TRAIL-induced apoptosis in certain cell types. ARAP1 also affects cell spreading.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ARAP1
Applications Immunoprecipitation, Western Blot, Immunocytochemistry
Classification Monoclonal
Clone ARAP1-2
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 15mM sodium azide; pH 7.4
Gene Arap1
Gene Accession No. Q96P48
Gene Alias 2410002L19Rik; arap1; Arf-GAP with Rho-GAP domain, ANK repeat and PH domain-containing protein 1; ArfGAP with RhoGAP domain, ankyrin repeat and PH domain 1; ARF-GAP, RHO-GAP, ankyrin repeat, and pleckstrin homology domains-containing protein 1; centaurin, delta 2; centaurin-delta-2; Centd2; cnt-d2; Kiaa0782; mKIAA0782
Gene Symbols Arap1
Host Species Mouse
Immunogen His6-tagged protein corresponding to amino acids 1190-1450 of human ARAP1.
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 116985
Target Species Human
Content And Storage 4°C, do not freeze
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.