Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ARAP2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA523326
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA523326 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA523326 Supplier Invitrogen™ Supplier No. PA523326
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The amino acid sequence used as immunogen is 87% homologous in Mouse and Rat, 81% homologous in Canine.

ARAP2 also known as Centaurin-delta-1 is a protein with ARF-GAP, RHO-GAP, ankyrin repeat, RAS-associating, and pleckstrin homology domains. A phosphatidylinositol-3, 4, 5-trisphosphate-dependent GTPase-activating protein, ARAP2, binds to RhoA-GTP, but lacks the predicted catalytic arginine in the RHO-GAP domain and does not have RHO-GAP activity. It modulates actin cytoskeleton remodeling by regulating ARF and RHO family members. Usually ARAP2 is activated by phosphatidylinositol-3, 4, 5-trisphosphate (PtdIns(3, 4, 5)P3) binding and can be activated by phosphatidylinositol-3, 4-bisphosphate (PtdIns(3, 4, 5)P2) binding, albeit with lower efficiency. ARAP2 associates with focal adhesions and functions downstream of RhoA to regulate focal adhesion dynamics. It is usually detected in brain, thymus, lymph node, thyroid, spinal cord, trachea, heart, skeletal muscle, spleen, kidney, liver, placenta, lung and peripheral blood leukocytes.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ARAP2
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% BSA and 0.05% sodium azide
Gene ARAP2
Gene Accession No. Q8WZ64
Gene Alias Arap2; Arf and Rho GAP adapter protein 2; arf-GAP with Rho-GAP domain, ANK repeat and PH domain-containing protein 2; ArfGAP with RhoGAP domain, ankyrin repeat and PH domain 2; centaurin; centaurin, delta 1; centaurin-delta-1; CENTD1; Cnt-d1; Gm148; KIAA0580; mKIAA0580; PARX; PARX protein; Protein PARX
Gene Symbols ARAP2
Host Species Rabbit
Immunogen A portion of amino acids 1670-1704 of human ARAP2.
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 116984
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.