Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ARAP3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA560158
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA560158 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA560158 Supplier Invitrogen™ Supplier No. PA560158
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MPNSIYFGLD SRGRAQAAQD KAPDSSQISA PTPALRPTTG TVHIMDPGCL YYGVQPVGTP GAPDRRESRG VCQGRAEHRL SRQDLEARED AGYASLELPG DSTLLSPTLE TEETSDDLIS PYASFSF Highest antigen sequence identity to the following orthologs: Mouse - 80%, Rat - 80%.

Ubiquitin is a conserved 76 amino acid polypeptide and can affect proteasomal degradation of the protein it is bound to, or mediate interactions with other proteins related to post-translational modifications. The degradation of cellular regulatory proteins by the Uubiquitin pathway is important as it controls the cellular growth and proliferation. Ubiquitin-dependent proteolysis occurs after a covalent attachment of the peptide to a lysine residue of a protein, which involves three enzymatic reactions: E1, E2 and E3. The first reaction involves ubiquitin-activating enzyme. The third reaction uses enzyme ubiquitin ligase (E3) to transfer the activated ubiquitin from E2 to a lysine residue on a protein, or directly transfers the ubiquitin from E2 to the substrate.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ARAP3
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.4 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ARAP3
Gene Accession No. Q8WWN8
Gene Alias ARAP3; Arf and Rho GAP adapter protein 3; arf-GAP with Rho-GAP domain, ANK repeat and PH domain-containing protein 3; ArfGAP with RhoGAP domain, ankyrin repeat and PH domain 3; ARF-GAP, RHO-GAP, ankyrin repeat and plekstrin homology domains-containing protein 3; centaurin; centaurin, delta 3; centaurin-delta-3; CENTD3; Cnt-d3; DRAG1; FLJ21065; phosphoinositide binding protein; PtdIns(3,4,5)P3-binding protein
Gene Symbols ARAP3
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ARAP3. Recombinant protein control fragment (Product #RP-90738).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64411
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.