Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ARC Mouse anti-Human, Mouse, Porcine, Rat, Clone: 1G4B5, Proteintech
SDP

Catalog No. 501736970 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-6970 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-6970 Supplier Proteintech Group Inc Supplier No. 665501IG150UL
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Required for consolidation of synaptic plasticity as well as formation of long-term memory. Regulates endocytosis of AMPA receptors in response to synaptic activity. Required for homeostatic synaptic scaling of AMPA receptors. Localized in: Cytoplasm, cytoskeleton. Endosome. Acrosome. Cell junction, synapse, postsynaptic cell membrane, postsynaptic density. Cell projection, dendrite. Cell projection, dendritic spine. Cell junction, synapse. Associated with the cell cortex of neuronal soma and dendrites. Enriched in postsynaptic density of dendritic spines. Associated with the sperm tail.

Specifications

Antigen ARC
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 1G4B5
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene ARC
Gene Accession No. Q63053, Q7LC44, Q9WV31
Gene Alias ARC, ARC/ARG3.1, Arg3.1, KIAA0278
Gene Symbols Arc
Host Species Mouse
Immunogen ARC Fusion Protein Ag9445
Purification Method Protein G
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 110260202, 11838, 23237, 54323
Target Species Human, Mouse, Pig, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.