Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ARC/NOL3 Antibody - BSA Free, Novus Biologicals™
SDP

Catalog No. p-7199899 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB018196 0.025 mg
NB500257 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB018196 Supplier Novus Biologicals Supplier No. NB500257SS
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ARC/NOL3 Polyclonal specifically detects ARC/NOL3 in Human samples. It is validated for Western Blot.

Specifications

Antigen ARC/NOL3
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000
Formulation PBS with 0.02% Sodium Azide
Gene Accession No. O60936
Gene Alias Apoptosis repressor with CARD, ARCFLJ35304, CARD2, Muscle-enriched cytoplasmic protein, MYC, MYP, NOP30, NOPnucleolar protein 3, nucleolar protein 3 (apoptosis repressor with CARD domain), Nucleolar protein of 30 kDa
Gene Symbols NOL3
Host Species Rabbit
Immunogen A synthetic peptide made to residues 125-141 (PELQTLTRPGALRAPRR) of the human Nop30 protein.
Molecular Weight of Antigen 33 kDa
Purification Method Affinity Purified
Quantity 0.025 mg
Regulatory Status RUO
Research Discipline Apoptosis
Primary or Secondary Primary
Gene ID (Entrez) 8996
Test Specificity This is specific for the human Nop30 protein (Gene names - ARC, NOP or NOL3).
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.