Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ARC Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5120251
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5120251 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5120251 Supplier Invitrogen™ Supplier No. PA5120251
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Positive test controls include: SW620, Mouse heart. The target is usually found in the following locations: Cytoplasm, Lipid-anchor, Membrane, Mitochondrion, Nucleus, Sarcoplasmic reticulum, nucleolus. Immunogen sequence: MGNAQERPSE TIDRERKRLV ETLQADSGLL LDALLARGVL TGPEYEALDA LPDAERRVRR LLLLVQGKGE AACQELLRCA QRTAGAPDPA WDWQHVGPGY RDRSYDPPCP GHWTPEAPGS GTTCPGLPRA SDPDEAGGPE GSEAVQSGTP EEPEPELEAE ASKEAEPEPE PEPELEPEAE AEPEPELEPE PDPEPEPDFE ERDESEDS.

ARC (also known as NOL3 - Nucleolar protein 3) has two isoforms. Isoform 1 is involved in RNA splicing. Isoform 2 functions as an apoptosis repressor that blocks multiple modes of cell death. ARC inhibits extrinsic apoptotic pathways two different ways. The first is by interacting with FAS and FADD upon FAS activation blocking death-inducing signaling complex (DISC) assembly. The second way is by interacting with CASP8 in a mitochondria localization-and phosphorylation-dependent manner, which in turn limits the amount of soluble CASP8 avaliable for DISC-mediated activation. ARC also inhibits intrinsic apoptotic pathways in response to a wide range of stresses through interactions with BAX. This results in BAX inactivation and prevents mitochondrial dysfunction and release of pro-apoptotic factors. Mutations affecting the gene can results in myoclonus, familiam cortical.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ARC
Applications ELISA, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.48 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene NOL3
Gene Accession No. O60936, Q9D1X0
Gene Alias activity regulated cytoskeletal-associated protein; activity regulated cytoskeleton associated protein; activity-regulated cytoskeleton-associated protein; Activity-regulated gene 3.1 protein; activity-regulated gene 3.1 protein homolog; Apoptosis repressor with CARD; ARC; arc {ECO:0000312; ARC/ARG3.1; Arc3.1; Arg3.1; B430311C09Rik; C86064; FCM; hArc; KIAA0278; mArc; Muscle-enriched cytoplasmic protein; MYC; MYP; Nol3; NOP; Nop 30; Nop30; Nucleolar protein 3; nucleolar protein 3 (apoptosis repressor with CARD domain); nucleolar protein of 30 kDa; rg3.1; RGD:62037}
Gene Symbols NOL3
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-208 of human NOL3 (NP_003937.1).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 78688, 8996
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.