Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ARC Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA519906
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA519906 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA519906 Supplier Invitrogen™ Supplier No. PA519906
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

In Western blot applications, this antibody detects a band at ~25kDa. A suggested positive control is Hela cell lysate. PA5-19906 can be used with blocking peptide PEP-0032.

ARC (also known as NOL3 - Nucleolar protein 3) has two isoforms. Isoform 1 is involved in RNA splicing. Isoform 2 functions as an apoptosis repressor that blocks multiple modes of cell death. ARC inhibits extrinsic apoptotic pathways two different ways. The first is by interacting with FAS and FADD upon FAS activation blocking death-inducing signaling complex (DISC) assembly. The second way is by interacting with CASP8 in a mitochondria localization-and phosphorylation-dependent manner, which in turn limits the amount of soluble CASP8 avaliable for DISC-mediated activation. ARC also inhibits intrinsic apoptotic pathways in response to a wide range of stresses through interactions with BAX. This results in BAX inactivation and prevents mitochondrial dysfunction and release of pro-apoptotic factors. Mutations affecting the gene can results in myoclonus, familiam cortical.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ARC
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene NOL3
Gene Accession No. O60936
Gene Alias activity regulated cytoskeletal-associated protein; activity regulated cytoskeleton associated protein; activity-regulated cytoskeleton-associated protein; Activity-regulated gene 3.1 protein; activity-regulated gene 3.1 protein homolog; Apoptosis repressor with CARD; ARC; arc {ECO:0000312; ARC/ARG3.1; Arc3.1; Arg3.1; B430311C09Rik; C86064; FCM; hArc; KIAA0278; mArc; Muscle-enriched cytoplasmic protein; MYC; MYP; Nol3; NOP; Nop 30; Nop30; Nucleolar protein 3; nucleolar protein 3 (apoptosis repressor with CARD domain); nucleolar protein of 30 kDa; rg3.1; RGD:62037}
Gene Symbols NOL3
Host Species Rabbit
Immunogen A peptide corresponding to amino acids 191 to 208 of human ARC.
Purification Method Antigen Affinity Chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8996
Target Species Human
Content And Storage Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.