Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ARCA Polyclonal Antibody
GREENER_CHOICE

Catalog No. FEPA5144460
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEPA5144460 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEPA5144460 Supplier Invitrogen™ Supplier No. PA5144460
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ARCA Polyclonal Antibody for Western Blot, ELISA

Member of the two-component regulatory system ArcB/ArcA. Represses a wide variety of aerobic enzymes under anaerobic conditions. Controls the resistance of E.coli to dyes; required for expression of the alkaline phosphatase and sex factor F genes; it may also be involved in the osmoregulation of envelope proteins. When activated by ArcB, it negatively regulates the expression of genes of aerobic function. Activates the transcription of the plfB operon by binding to its promoter.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ARCA
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 2.77 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% Proclin 300; pH 7.4
Gene arcA
Gene Accession No. P0A9Q1, P23793
Gene Alias cpxC; dye; fexA; msp; seg; sfrA
Gene Symbols arcA
Host Species Rabbit
Immunogen Recombinant Escherichia coli (strain K12) ARCA protein.
Purification Method Protein A/G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 69703137, 82262143
Target Species Bacteria
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.