Learn More
National Safety Apparel™ Enespro™ ArcGuard™ 40 cal Contractor Arc Flash Kit with Short Coat and Bib Overall (08)
SDP
40 cal Arc flash kit with short coat and bib overalls with gloves
$1881.00 - $2060.00
Specifications
Ankle Style Open
Arc Flash Rating 40 cal/cm²
Certifications/Compliance CAT 4, NFPA 70E, CSA Z462, ASTM F1506, OSHA 1910.269
Closure Type FR Hook and Loop
Color Tan
View More Specs
Products 8
Catalog Number Mfr. No. Size Price Quantity & Availability  
Catalog Number Mfr. No. Size Price Quantity & Availability  
17-001-1662
SDP
View Documents
National Safety Apparel™
KIT4SC40ECSM08
Small
Each for $1,881.00
Only null left
Add to Cart
17-001-1667
SDP
View Documents
National Safety Apparel™
KIT4SC40ECMD08
Medium
Each for $1,881.00
Only null left
Add to Cart
17-001-1672
SDP
View Documents
National Safety Apparel™
KIT4SC40ECLG08
Large
Each for $1,881.00
Only null left
Add to Cart
17-001-1677
SDP
View Documents
National Safety Apparel™
KIT4SC40ECXL08
X-Large
Each for $1,881.00
Only null left
Add to Cart
17-001-1682
SDP
View Documents
National Safety Apparel™
KIT4SC40EC2X08
2X-Large
Each for $1,936.00
Only null left
Add to Cart
17-001-1687
SDP
View Documents
National Safety Apparel™
KIT4SC40EC3X08
3X-Large
Each for $1,936.00
Only null left
Add to Cart
17-001-1692
SDP
View Documents
National Safety Apparel™
KIT4SC40EC4X08
4X-Large
Each for $1,996.00
Only null left
Add to Cart
17-001-1697
SDP
View Documents
National Safety Apparel™
KIT4SC40EC5X08
5X-Large
Each for $2,060.00
Only null left
Add to Cart
Description
Description
  • ArcGuard™ Economy™ Short Coat and Bib Overall
  • 14 in. Class 2 Rubber Insulating Gloves
  • 12 in. Leather Glove Protectors
  • Arc Flash Hood Unit with faceshield & hard hat
  • Safety Glasses
  • Hood bag
  • Glove bag Gear bag
Specifications
Specifications
Open
CAT 4, NFPA 70E, CSA Z462, ASTM F1506, OSHA 1910.269
Tan
Arc Flash Kit
Adjustable
Unisex
40 cal/cm²
FR Hook and Loop
Cotton, Nylon
Stitched
Utilities, Manufacturing, Electrical Contractors
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.