Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

National Safety Apparel™ Enespro™ AGP 40 cal Arc Flash Jacket
SDP

Catalog No. p-7235733
Click to view available options
Size:
X-Small
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large

AGP Arc Flash Jacket, 40 Cal, CAT 4, Back Stretch Panels, Drawstring Waist, Pleated Elbows, Knit Lined Collar

  • Single-layer 40 cal arc flash protection
  • Up to 20% lighter, 48% thinner, and 56% more flexible than other premium arc flash suits
  • Using GORE™ PYRAD™ comfort science to keep the wearer cool, dry, comfortable, and unrestricted while working
  • FR cuffs improve fit with electrical gloves
  • Covered zip front closure with large pull tab
  • Ergonomic design with added stretch back panels for ease of movement and flexibility
  • Lined collar for soft touch and moisture-wicking
  • Enhanced visibility with 3M™ Scotchlite™ FR silver segmented reflective trim

Specifications

Size X-Small
Gender Unisex
Material ePTFE, Polyamide, FR Viscose, Aramid, Carbon
Color Navy
Closure Type Zipper
Arc Flash Rating 40 cal/cm²
Certifications/Compliance CAT 4, NFPA 70E, CSA Z462, OSHA 1910.269, ASTM F1506
Product Type Jacket
For Use With (Application) Utilities, Electrical Contractors, Manufacturing
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.