Learn More
Invitrogen™ ARCH Monoclonal Antibody (1G4)

Description
Peptide Sequence: MAQEEEDVRD YNLTEEQKAI KAKYPPVNRK YEYLDHTADV QLHAWGDTLE EAFEQCAMAM FGYMTDTGTV EPLQTVEVET QGDDLQSLLF HFLDEWLYKF SADEFFIPRV GRRIFIVQAP SGNRSQSNNI FSNAGL.
Specifications
Specifications
| Antigen | ARCH |
| Applications | ELISA, Western Blot |
| Classification | Monoclonal |
| Clone | 1G4 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with no preservative; pH 7.4 |
| Gene | ZBTB8OS |
| Gene Accession No. | Q8IWT0 |
| Gene Alias | ARCH; ARCH2; archease (ARCH); archease-like protein; protein archease; Protein ZBTB8OS; RGD1562249; ZBTB8OS; Zbtb8os-ps1; zinc finger and BTB domain containing 8 opposite strand; zinc finger and BTB domain containing 8 opposite strand, pseudogene 1; zinc finger and BTB domain-containing opposite strand protein 8 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.