Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ARCH Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA561213
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA561213 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA561213 Supplier Invitrogen™ Supplier No. PA561213
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: EEDVRDYNLT EEQKAIKAKY PPVNR Highest antigen sequence identity to the following orthologs: Mouse - 92%, Rat - 96%.

Component of the tRNA-splicing ligase complex required to facilitate the enzymatic turnover of catalytic subunit RTCB. Together with DDX1, acts by facilitating the guanylylation of RTCB, a key intermediate step in tRNA ligation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ARCH
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ZBTB8OS
Gene Accession No. Q8IWT0
Gene Alias ARCH; ARCH2; archease (ARCH); archease-like protein; protein archease; Protein ZBTB8OS; RGD1562249; ZBTB8OS; Zbtb8os-ps1; zinc finger and BTB domain containing 8 opposite strand; zinc finger and BTB domain containing 8 opposite strand, pseudogene 1; zinc finger and BTB domain-containing opposite strand protein 8
Gene Symbols ZBTB8OS
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ARCH. Recombinant protein control fragment (Product #RP-98927).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 339487
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.