Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

archaemetzincin-1 Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody has been used in 1 publication

Supplier:  Novus Biologicals NBP191677

 View more versions of this product

Catalog No. NBP191677


Only null left
Add to Cart

Description

Description

archaemetzincin-1 Polyclonal specifically detects archaemetzincin-1 in Human samples. It is validated for Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Electron Microscopy.
Specifications

Specifications

archaemetzincin-1
Polyclonal
Flow Cytometry, ELISA, Immunohistochemistry 1:20 - 1:50, Immunohistochemistry-Paraffin 1:20 - 1:50, Electron Microscopy
Q400G9
AMZ1
This antibody was developed against Recombinant Protein corresponding to amino acids:AIKEHERWLAMCIQALQREVAEEDLVQVDRAVDALDRWEMFTGQLPATRQDPPSSRDSVGLRKVLGDKFSSLRRKLSARKLARAESAPRPWDG
0.1 mL
Primary
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoassay
Unconjugated
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide
archaelysin family metallopeptidase 1
Rabbit
Affinity Purified
RUO
155185
Human
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.

For Research Use Only