Learn More
DR Instruments Archaeology Probe Kit for Exploration
SDPSCI_ED
Catalog No. s72151
Click to view available options
:
Archaeology Probe Kit; Includes: 12 Probes
California Proposition 65 WARNING: Cancer and Reproductive Harm
Catalog No. S72151
Supplier: DR Instruments DRAK60
Only null left
Add to Cart
Add to Cart
California Proposition 65 WARNING: Cancer and Reproductive Harm
Archaeology Probe Kit offers metal rods or tubes that are pushed into unexcavated deposits to locate as yet unexposed hard features such as walls, floors, bedrock, bone, etc.
  • This collection provides a diverse selection of probes essential for archaeological endeavors.
  • These instruments comprise metal rods or tubes employed to penetrate unexcavated deposits, facilitating the discovery of concealed elements like walls, floors, bedrock, and bone.
Specifications
Product Type Archaeology Probe
Material Stainless Steel
For Use With (Application) Archaeological activities
Includes 12 Probes
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.