Learn More
GSC Go Science Crazy Archimedes Principle Kit
SCI_ED
Catalog No. s28116 Shop All GSC Products
Encompass
Change view
Click to view available options
:
Archimedes Principle Kit
Catalog No.
S28116 Archimedes Principle Kit
1 options
Not a toy. Product intended for ages 13 and up.
Catalog No. S28116
Supplier: GSC Go Science Crazy ARCHKT
Only null left
Add to Cart
Add to Cart
Not a toy. Product intended for ages 13 and up.
Classroom apparatus that helps teach students about Archimedes Principle and buoyancy. Objects appear to weigh less when submerged in water because the objects are buoyed up by a force equal to the weight of the fluid displaced.
  • Teaches about laws of buoyancy
  • Includes hollow solid cylinder, instructions
Specifications
Product Type Archimedes Principle Kit
For Use With (Application) To learn about Archimedes principle and buoyancy laws
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov