Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abbott ARCHITECT B·R·A·H·M·S PCT Test

Catalog No. p-8349038
Click to view available options
Quantity:
100 Tests
This item is not returnable. View return policy
This item is not returnable. View return policy

Used in conjunction with other laboratory findings and clinical
assessments, the ARCHITECT B∙R∙A∙H∙M∙S PCT assay is intended for use
as an:

  • Aid in the risk assessment of critically ill patients on their first day of intensive care unit (ICU) admission for progression to severe sepsis and septic shock
  • Aid in assessing the cumulative 28-day risk of all-cause mortality for patients diagnosed with severe sepsis or septic shock in the ICU or when obtained in the emergency department or other medical wards
    prior to ICU admission, using a change in PCT level over time
  • Aid in decision making on antibiotic therapy for patients with suspected or confirmed lower respiratory tract infections (LRTI) – defined as community-acquired pneumonia (CAP), acute bronchitis, and acute exacerbation of chronic obstructive pulmonary disease (AECOPD) – in an inpatient setting or an emergency department
  • Aid in decision making on antibiotic discontinuation for patients with
    suspected or confirmed sepsis.

Reagents:

Microparticles: Tris-based buffer with protein (bovine) stabilizer, rat IgG, and Triton X-405. Minimum concentration: 0.06% solids. Preservatives: ProClin 950 and sodium azide.

Conjugate: Anti-PCT (mouse, monoclonal) acridinium-labeled conjugate in phosphate buffer with protein (bovine) stabilizer and Triton X-405. Minimum concentration: 270ng/mL. Preservatives: ProClin 950 and sodium azide.

Specifications

Content And Storage 2°C to 8°C Do not freeze
Sample Type Serum, plasma
Type PCT Reagent Kit
For Use With (Application) Quantitative determination of PCT (Procalcitonin) in human serum and plasma on the automated ARCHITECT analyzer
Includes Microparticles: 1 x 8.6mL; Conjugate: 1 x 5.9mL
pH At 20 °C (68 °F): Microparticles: 7; Conjugate: 6.5
Quantity 100 Tests

For In Vitro Diagnostic Use

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.