Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abbott ARCHITECT B·R·A·H·M·S PCT Calibrator Kit

Catalog No. 06p2201a
Click to view available options
Quantity:
6 Bottles
This item is not returnable. View return policy
This item is not returnable. View return policy

For the calibration of the ARCHITECT iSystem when used for the quantitative determination of procalcitonin (PCT) in human serum and plasma.

6 Bottles (2.0mL each) of ARCHITECT B·R·A·H·M·S PCT Calibrators A, B–F.

  • Calibrator A contains normal human plasma. Preservatives: ProClin 950 and sodium azide
  • Calibrators B–F contain different concentrations of recombinant PCT in phosphate buffer.
    Preservatives: ProClin 950 and sodium azide.

Specifications

Content And Storage Frozen at −10°C or colder when not in use.
Concentration CAL A: 0.00 μg/L, CAL B: 0.10 μg/L. CAL C:0.50 μg/L, CAL D: 12.10 μg/L, CAL E: 20.50 μg/L, CAL F: 100.00 μg/L
Type PCT Calibration Kit
Includes 6 Bottles (2.0mL each) of ARCHITECT B·R·A·H·M·S PCT Calibrators (A, B to F)
Intended Use In Vitro Diagnostic Use
Packaging Bottle
Quantity 6 Bottles
Form Liquid

For In Vitro Diagnostic Use

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.