Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ARCN1 Mouse anti-Canine, Human, Rat, Clone: OTI4H9, liquid, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene TA504314S

Description
This gene maps in a region, which include the mixed lineage leukemia and Friend leukemia virus integration 1 genes, where multiple disease-associated chromosome translocations occur. It is an intracellular protein. Archain sequences are well conserved among eukaryotes and this protein may play a fundamental role in eukaryotic cell biology. It has similarities to heat shock proteins and clathrin-associated proteins, and may be involved in vesicle structure or trafficking.Specifications
ARCN1 | |
Monoclonal | |
0.92 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
P48444, Q66H80 | |
ARCN1 | |
Full length human recombit protein of human ARCN1 produced in HEK293T cell. | |
30 μL | |
Primary | |
Canine, Human, Rat | |
Antibody | |
IgG1 |
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
OTI4H9 | |
Unconjugated | |
ARCN1 | |
COPD, SRMMD | |
Mouse | |
Affinity Chromatography | |
RUO | |
300674, 372, 479415 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction