Learn More
Epredia™ Arcos™ Trays and Storage
Catalog No. p-6817022 Shop All Epredia Products
Change view
Click to view available options
Quantity:
1
25 Pk.
6 Pk.
Product Type:
Cardboard Storage Box
Hook
Slide Tray
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Product Type
B84710001 6 Pk. Slide Tray
B84710005 25 Pk. Cardboard Storage Box
B84710006 1 Hook
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. B84710001 Supplier Epredia™ Supplier No. B84710001
Enhance traceability and organization with trays, storage cabinets and cardboard sleeves for the Epredia™ Arcos™ block management system.
Smart and Secure Storage
  • The Arcos system offers flexible archiving solutions to fit customer needs for both short and long-term archiving. The Arcos storage cabinets offer two configurations with 18 and 24 tray capacity. These are designed specifically for high-density storage that can be stacked to optimize capacity and space.
  • For short-term archiving, the cabinets can be placed in the main lab area so the blocks can be accessed with ease. The 18 tray cabinets can be stacked three-units high, and the 24 tray cabinets can be stacked two-units high.
  • For long-term archiving, cardboard sleeves may be used as an alternative storage option in place of the Arcos cabinets. Each sleeve can hold one Arcos tray, up to 240 blocks.
Specifications
Product Type Slide Tray
Product Line Epredia Arcos
Quantity 6 Pk.
Epredia™ Arcos™ instrumentation requires the use of Arcos trays, sold separately. Arcos cabinets are highly recommended for storage.