Learn More
Thermo Scientific™ GENESYS™ 30 Visible Spectrophotometer
Catalog No. 13875125
Click to view available options
Includes:
GENESYS 30 Spectrophotometer
Performance Verification Kit
Warranty:
2 yrs std + 1 yr ext upon reg
Standard
This item is not returnable. View return policy
This item is not returnable. View return policy
Perform both basic and complex analyses with the Thermo Scientific™ GENESYS™ 30 Visible Spectrophotometer, an intuitive, reliable solution for education or in the lab.
Get high precision and functionality in a simple yet sophisticated visible spectrophotometer. The Thermo Scientific™ GENESYS™ 30 Visible Spectrophotometer performs both basic and complex analyses for educational or industrial use. Its intuitive design allows students and teachers to focus on scientific data and concepts rather than on learning how to use the instrument. In the lab, technicians of all levels working in routine QA/QC applications can easily obtain credible results to confirm quality or inform next steps. Results can be reported quickly as digital or printed data.
Use with the PV application in the Settings menu of the GENESYS 30 to perform the fundamental PV tests to verify that the instrument is functioning correctly.
TRUSTED_SUSTAINABILITY
Specifications
Includes Performance Verification Kit
Warranty Standard
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.