Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Radians Arctic Radwear™ Cooling Towel

Click to view available options
Color:
Hi-Vis Yellow
Description
- For hot outdoor activities such as construction work, tough workouts or strenuous outdoor/indoor activity
- Soak for 2 to 3 min and wear for up to 5 hrs. of cooling relief (depending on humidity)
- Reusable packaging design
- Safe and reusable
- Jobsite proven
- Anti-microbial treated to help prevent mold build-up and unwanted smells
- Heat stress safety management
Specifications
Specifications
| Product Type | Cooling Towel |
| Dimensions (L x W) | 26 x 17 in. |
| For Use With (Application) | For hot outdoor activities such as construction work, tough workouts or any strenuous outdoor or indoor activity |
| Quantity | 10 Pk., 50/Cs. |
Warning: Cancer and reproductive harm
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction