Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Radians Arctic Radwear™ Hard Hat Cooling Sweatband
SDP

Catalog No. p-7221215
Click to view available options
Quantity:
10 Pk., 50/Cs.
100 Pk., 1000/Cs.
Product Type:
Hard Hat Cooling Sweatband

Accelerates the evaporative cooling process

  • Easy installation
  • Safe and reusable
  • Job-site proven
  • Anti-microbial treated to help prevent mold build up and unwanted smells
  • Heat stress safety management- cools body temp
  • Lasts up to 5 hrs. depending on humidity levels
  • Designed to keep cool during hot outdoor activities like construction work, tough workouts or strenuous outdoor and indoor activity

How to Use:

  • Soak in water for 2 to 3 min.
  • Wave in air for 10 to 15 sec. to activate the material
  • Securely snap sweatband over existing hard hat suspension headband
  • Reactivate in extreme heat or humid conditions by repeating process

Notes:

  • Hand wash only with mild detergent
  • Do NOT bleach
  • Hard hat and suspension sold separately

Specifications

Color Blue
For Use With (Application) For hot outdoor activities like construction work
Quantity 10 Pk., 50/Cs.

Warning: Cancer and reproductive harm

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.