Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Radians Arctic Radwear™ Headband

Click to view available options
Color:
Blue
Blue Paisley
Hi-Vis Yellow
Red Paisley
Description
- Safe and reusable
- Jobsite proven
- Anti-microbial treated to help prevent mold build-up and unwanted smells
- Heat stress safety management
- One size fits all
- Soak the band in water for 2 to 3 min. and wear for up to five hours
- Stretch-fit spandex design allows the band to move with the body
- Allows for optimum blood circulation
- Designed to keep cool during hot outdoor activities like construction work, tough workouts or strenuous outdoor and indoor activity
Specifications
Specifications
| Product Type | Headband |
| For Use With (Application) | For hot outdoor activities such as construction work, tough workouts or any strenuous outdoor or indoor activity |
| Quantity | 10 Pk., 50/Cs. |
Warning: Cancer and reproductive harm
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction