Learn More
Heathrow Scientific™ Arctic Squares™ Cryogenic Tube Storage Box
GREENER_CHOICE
Catalog No. 17100427
Change view
Click to view available options
Capacity (Metric):
100 Cryogenic Vials
25 Cryogenic Vials
81 Cryogenic Vials
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
17-100-427 81 Cryogenic Vials
17-100-462 100 Cryogenic Vials
17-100-446 25 Cryogenic Vials
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 17-100-427 Supplier Heathrow Scientific™ Supplier No. HS23381B
Add to Cart
Add to Cart
Safely store cryogenic vials in this Arctic Squares™ Cryogenic Tube Storage Box.
  • Our polycarbonate boxes are designed with unique features to help orient, identify and access vials with ease
  • Alphanumeric labeling on lid or base (depending on the unit) for quick location and retrieval of tubes
  • Liquid nitrogen vent and drainage holes
  • Magnet on top of forceps enables easy storing on metal surfaces
  • Stackable, saves valuable space in the lab
Specifications
Capacity (Metric) 81 Cryogenic Vials
Closure Type Slide On Lid
Color Assorted
Material Polycarbonate
Shape Square
Type Cryo Box
Holds 2.0 mL Cryogenic Vials
Includes Forceps
Tube Array 9 x 9
Dimensions (L x W x H) 5.2 x 5.2 x 2 in. (13.3 x 13.3 x 5.3 cm)