Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ARD1A Monoclonal Antibody (06), Invitrogen™
Mouse Monoclonal Antibody
$285.50 - $493.00
Specifications
Antigen | ARD1A |
---|---|
Clone | 06 |
Concentration | 1 mg/mL |
Applications | ELISA |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
PIMA543486
![]() |
Thermo Scientific
MA543486 |
50 μL |
Each for $285.50
|
|
|||||
PIMA543042
![]() |
Thermo Scientific
MA543042 |
100 μL |
Each for $493.00
|
|
|||||
Description
Specificity: Human ARD1
N-alpha-acetylation is one of the most common protein modifications that occurs during protein synthesis and involves the transfer of an acetyl group from acetyl-coenzyme A to the protein alpha-amino group. ARD1A, together with NATH.Specifications
ARD1A | |
1 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with no preservative | |
2310039H09Rik; alpha-N-acetyltransferase 1A; ARD1; ARD1 homolog A, N-acetyltransferase; Ard1a; ARD1P; DXS707; hARD1; MCOPS1; N(alpha)-acetyltransferase 10, NatA catalytic subunit; N(alpha)-acetyltransferase 10, NatA catalytic subunitNalpha acetyltransferase 10; Naa10; N-acetyltransferase ARD1 homolog; N-acetyltransferase ARD1, human homolog of; Nalpha acetyltransferase 10; N-alpha-acetyltransferase 10; N-alpha-acetyltransferase 10, NatA catalytic subunit; natA catalytic subunit Naa10; NATD; N-terminal acetyltransferase complex ARD1 subunit homolog A; OGDNS; RGD1565315; Te2 | |
NAA10 | |
IgG1 | |
Protein A | |
Antibody |
06 | |
ELISA | |
Unconjugated | |
Mouse | |
Human | |
P41227 | |
8260 | |
Recombinant Human ARD1 protein. | |
Primary | |
-20° C, Avoid Freeze/Thaw Cycles | |
NAA10 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title