Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ARF1 Antibody (1A9/5), HRP, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP308340H
Description
ARF1 Monoclonal antibody specifically detects ARF1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry (Paraffin)Specifications
ARF1 | |
Monoclonal | |
HRP | |
ADP-ribosylation factor 1 | |
C-terminal human ARF1 peptide SNQLRNQ (Uniprot: P84077) | |
100 μg | |
Signal Transduction | |
375 | |
Store at 4°C in the dark. | |
IgG2a κ |
Western Blot, Immunohistochemistry (Paraffin) | |
1A9/5 | |
PBS | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction