Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ARF1 Antibody (1B3), Alexa Fluor™ 594, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP197935AF594
Description
ARF1 Monoclonal specifically detects ARF1 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence.Specifications
ARF1 | |
Monoclonal | |
Alexa Fluor 594 | |
50 mM sodium borate with 0.05% sodium azide | |
ARF1 | |
Recombinant human ARF1 (1-181aa) purified from E. coli | |
100 μL | |
375.0 | |
Store at 4°C in the dark. |
Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence | |
1B3 | |
Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence | |
ADP-ribosylation factor 1 | |
Mouse | |
Protein G purified | |
Primary | |
Human | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction