Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ARF1 Antibody (ARFS 1A9/5), FITC, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB1002635F
Description
ARF1 Monoclonal specifically detects ARF1 in Human samples. It is validated for ELISA, Immunofluorescence.Specifications
ARF1 | |
Monoclonal | |
FITC | |
PBS with 0.05% Sodium Azide | |
ARF1 | |
C-terminal Human ARF1 peptide SNQLRNQ | |
0.1 mL | |
Primary | |
ARFS 1A9/5 is Class 1 ARF specific. | |
Store at 4C in the dark. | |
IgG2a |
ELISA, Immunofluorescence | |
ARFS 1A9/5 | |
ELISA | |
ADP-ribosylation factor 1 | |
Mouse | |
Protein A purified | |
RUO | |
375 | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction