Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ARF1/ARF2/ARF3/ARF4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA518627
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA518627 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA518627 Supplier Invitrogen™ Supplier No. PA518627
Add to Cart
Add to Cart

Goat Polyclonal Antibody

This antibody is predicted to react with canine, mouse and rat based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 8,000.

ADP-ribosylation factor 1 (ARF1) is a member of the humanARFgene family. The family members encode small guanine nucleotide-binding proteins that stimulate the ADP-ribosyltransferase activity of cholera toxin and play a role in vesicular trafficking as activators of phospholipase D. The gene products, including 6ARFproteins and 11ARF-like proteins, constitute a family of the RAS superfamily. TheARFproteins are categorized as class I (ARF1,ARF2 andARF3), class II (ARF4 andARF5) and class III (ARF6), and members of each class share a common gene organization. TheARF1 protein is localized to the Golgi apparatus and has a central role in intra-Golgi transport. Multiple alternatively spliced transcript variants encoding the same protein have been found for this gene. [provided by RefSeq, Jul 2008].
TRUSTED_SUSTAINABILITY

Specifications

Antigen ARF1/ARF2/ARF3/ARF4
Applications Flow Cytometry, Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
Gene Arf4
Gene Accession No. P18085, P61204, P61205, P61206, P61750, P61751, P62330, P62331, P62332, P84077, P84078, P84079, P84082, P84083, P84084, P84085, Q8BSL7
Gene Alias 5430400P17Rik; AA407803; Ac1-253; ADP ribosylation factor 1; ADP ribosylation factor 3; ADP ribosylation factor 4; ADP ribosylation factor 5; ADP ribosylation factor 6; ADP-ribosylation factor 1; ADP-ribosylation factor 2; ADP-ribosylation factor 3; ADP-ribosylation factor 4; ADP-ribosylation factor 4-like protein; ADP-ribosylation factor 5; ADP-ribosylation factor 6; AI788669; AI854770; ARF; Arf1; Arf2; ARF3; ARF4; Arf5; ARF6; AW496366; I79_003112; Liver regeneration-related protein LRRG202; pan ARF; small GTP binding protein
Gene Symbols ARF1, Arf2, ARF3, Arf4, ARF5, ARF6
Host Species Goat
Immunogen Synthetic peptide sequence (EGLDWLSNQLRNQK) corresponding to the C-terminus amino acids of ARF1 (aa 168-181).
Purification Method Ammonium sulfate precipitation
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100856517, 11840, 11841, 11842, 11843, 11844, 11845, 140940, 375, 377, 378, 381, 382, 475206, 486562, 490679, 607073, 64310, 79117, 79119, 79120, 79121
Target Species Human, Mouse, Rat, Canine
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.