Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ARF1 Recombinant Rabbit Monoclonal Antibody (0C5B3)

Rabbit Recombinant Monoclonal Antibody
Supplier: Invitrogen™ MA535777
Description
Immunogen sequence: GQDKIRPLWR HYFQNTQGLI FVVDSNDRER VNEAREELMR MLAEDELRDA VLLVFANKQD LPNAMNAAEI TDKLGLHSLR H.
ADP-ribosylation factor 1 (ARF1) is a member of the human ARF gene family. The family members encode small guanine nucleotide-binding proteins that stimulate the ADP-ribosyltransferase activity of cholera toxin and play a role in vesicular trafficking as activators of phospholipase D. The gene products, including 6 ARF proteins and 11 ARF-like proteins, constitute a family of the RAS superfamily. The ARF proteins are categorized as class I (ARF1, ARF2 and ARF3), class II (ARF4 and ARF5) and class III (ARF6), and members of each class share a common gene organization. The ARF1 protein is localized to the Golgi apparatus and has a central role in intra-Golgi transport. Multiple alternatively spliced transcript variants encoding the same protein have been found for this gene.
Specifications
| ARF1 | |
| Recombinant Monoclonal | |
| 0.5 mg/mL | |
| PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P84077, P84078, P84079 | |
| ARF1 | |
| A synthetic peptide corresponding to a sequence within amino acids 70-150 of human ARF1 (P84077). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| ELISA, Western Blot, Immunocytochemistry | |
| 0C5B3 | |
| Unconjugated | |
| ARF1 | |
| ADP ribosylation factor 1; ADP ribosylation factor 1 L homeolog; ADP-ribosylation factor 1; Arf1; arf1.L; arf2; wu:fb33e02; wu:fb78h01; XELAEV_18043369mg | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 11840, 375, 64310 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction