Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ARF1 Mouse anti-Human, Mouse, Rat, Unconjugated, Clone: 1A9/5, Abnova™

Catalog No. 89337620 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-337-620 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-337-620 Supplier Abnova Corporation Supplier No. MAB21127
Add to Cart
Add to Cart

Mouse Monoclonal Antibody.

ARF1 Monoclonal antibody specifically detects ARF1 in Human, Mouse, Rat samples. It is validated for WB, IHC-P.

Specifications

Antigen ARF1
Applications Western Blot, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 1A9/5
Conjugate Unconjugated
Description ADP-ribosylation factor 1
Gene ARF1
Host Species Mouse
Immunogen A synthetic peptide corresponding to C-terminus of human ARF1 peptide
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) 375
Target Species Human, Mouse, Rat
Content And Storage Store at -20 to -80°C. Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.