Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ARF1 Rabbit anti-Human, Mouse, Rat, Polyclonal, FabGennix

Catalog No. 502206129
Encompass
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-220-6129 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-220-6129 Supplier Fabgennix Inc Supplier No. ARF1101AP
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ARF1 Polyclonal antibody specifically detects ARF1 in Human, Mouse, Rat samples. It is validated for Dot Blot, ELISA, Immunocytochemistry, Immunohistochemistry, Immunoprecipitation, Western Blot

Specifications

Antigen ARF1
Applications Dot Blot, ELISA, Immunocytochemistry, Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.5-1.5 mg/mL
Conjugate Unconjugated
Formulation proprietary buffer with 0.5% BSA, 30% glycerol and 0.02% sodium azide; pH 7.4-7.8
Gene ARF1
Gene Accession No. P84077, P84078, P84079
Gene Alias ADP ribosylation factor 1; ADP ribosylation factor 1 L homeolog; ADP-ribosylation factor 1; Arf1; arf1.L; arf2; wu:fb33e02; wu:fb78h01; XELAEV_18043369mg
Host Species Rabbit
Immunogen Synthetic peptide corresponding to unique amino acid sequence on Rat ARF1 protein. The peptide was covalently modified post-synthetically to achieve desied antigenicity.
Purification Method Affinity chromatography
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11840, 375, 64310
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.