Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ARF16 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5119707
This item is not returnable.
View return policy
Description
Positive Control: Rice tissue lysates. Subcellular Location: Nucleus.
Auxin response factors (ARFs) are transcriptional factors that bind specifically to the DNA sequence 5'-TGTCTC-3' found in the auxin-responsive promoter elements (AuxREs).
Specifications
| ARF16 | |
| Polyclonal | |
| Unconjugated | |
| ARF16 | |
| Auxin respoARF16nse factor 16 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 829131 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| ELISA, Western Blot | |
| 1 mg/mL | |
| TBS with 0.2% BSA, 50% glycerol and 0.05% sodium azide; pH 7.4 | |
| 0 | |
| ARF16 | |
| Synthetic peptide within Japonica rice aa 580-640. | |
| 100 μL | |
| Primary | |
| Plant | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction