Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ARF3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA529970
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA529970 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA529970 Supplier Invitrogen™ Supplier No. PA529970
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: Mouse brain. Predicted reactivity: Mouse (100%), Rat (100%), Xenopus laevis (99%), Pig (99%), Sheep (100%), Bovine (100%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

ADP-ribosylation factor 3 (ARF3) is a member of the human ARF gene family. These genes encode small guanine nucleotide-binding proteins that stimulate the ADP-ribosyltransferase activity of cholera toxin and play a role in vesicular trafficking and as activators of phospholipase D. The gene products include 6 ARF proteins and 11 ARF-like proteins and constitute 1 family of the RAS superfamily. The ARF proteins are categorized as class I (ARF1, ARF2, and ARF3), class II (ARF4 and ARF5) and class III (ARF6) and members of each class share a common gene organization. The ARF3 gene contains five exons and four introns.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ARF3
Applications Western Blot
Classification Polyclonal
Concentration 0.31 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7
Gene ARF3
Gene Accession No. P61204, P61205
Gene Alias 35-beta calcimedin; 5430400P17Rik; Ac1-253; ADP ribosylation factor 3; ADP-ribosylation factor 3; AI854770; Annexin A4; Annexin IV; Annexin-4; ANX4; ANXA4; Arf3; Chromobindin-4; Endonexin I; Lipocortin IV; Liver regeneration-related protein LRRG202; P32.5; PAP-II; PP4-X; small GTP binding protein
Gene Symbols ARF3
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human ARF3. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11842, 377
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.