Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ARF4 Polyclonal Antibody

ARF4 Polyclonal Antibody
Supplier: Thermo Scientific PA5109405
Description
ARF4 Polyclonal Antibody for Western Blot
Specifications
| ARF4 | |
| Polyclonal | |
| Unconjugated | |
| ARF4 | |
| AA407803; ADP ribosylation factor 4; ADP-ribosylation factor 2; ADP-ribosylation factor 4; ARF2; Arf4 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 11843, 378, 79120 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot, Immunocytochemistry | |
| 1.41 mg/mL | |
| PBS with 50% glycerol and 0.09% sodium azide; pH 7.3 | |
| P18085, P61750, P61751 | |
| ARF4 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-180 of human ARF4 (NP_001651.1). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction