Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ARF4L Antibody, Novus Biologicals™
SDP

Catalog No. NBP151927 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP151927 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP151927 Supplier Novus Biologicals Supplier No. NBP151927
Only null left
Add to Cart
Add to Cart

Goat Polyclonal Antibody

ARF4L Polyclonal specifically detects ARF4L in Human samples. It is validated for Western Blot, Peptide ELISA.

Specifications

Antigen ARF4L
Applications Western Blot, ELISA
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Western Blot 0.3 - 1 ug/ml, Peptide ELISA Detection limit 1:64000
Formulation Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide
Gene Accession No. P49703
Gene Alias ADP-ribosylation factor 4-like, ADP-ribosylation factor-like 4D, ADP-ribosylation factor-like protein 4D, ADP-ribosylation factor-like protein 4L, ARF4LADP-ribosylation factor-like 6, ARL6
Gene Symbols ARL4D
Host Species Goat
Immunogen Peptide with sequence C-KQDQPGALSAAEVEKR corresponding to internal region according to NP_001652.2.
Molecular Weight of Antigen 22 kDa
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 379
Test Specificity This antibody is expected not to cross-react to other ARL4 products.
Target Species Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.