Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ARF5 Antibody (SR1282), Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP32231450UL
Description
ARF5 Monoclonal antibody specifically detects ARF5 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow CytometrySpecifications
ARF5 | |
Monoclonal | |
Unconjugated | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Western Blot, Flow Cytometry | |
SR1282 | |
Western Blot 1:500-1:2000, Flow Cytometry 1:50 | |
ADP-ribosylation factor 5 | |
A synthesized peptide derived from human ARF5 (Uniprot #: P84085) | |
50 μg | |
Signal Transduction | |
381 | |
Store at -20° C. Avoid freeze/thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction