Learn More
Invitrogen™ ARF5 Monoclonal Antibody (1B6)

Description
Peptide Sequence: YFQNTQGLIF VVDSNDRERV QESADELQKM LQEDELRDAV LLVFANKQDM PNAMPVSELT DKLGLQHLRS RTWYVQATCA TQGTGLYDGL DWLSHELSKR.
Specifications
Specifications
| Antigen | ARF5 |
| Applications | ELISA, Western Blot |
| Classification | Monoclonal |
| Clone | 1B6 |
| Concentration | 0.2-1.0 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with no preservative; pH 7.4 |
| Gene | ARF5 |
| Gene Accession No. | P84085 |
| Gene Alias | ADP ribosylation factor 5; ADP-ribosylation factor 5; ARF5; EFL2; EPLG3; LERK3 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.