Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ARF5 Recombinant Rabbit Monoclonal Antibody (0U0D2)
GREENER_CHOICE

Catalog No. PIMA538062
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA538062 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA538062 Supplier Invitrogen™ Supplier No. MA538062
Add to Cart
Add to Cart

ARF5 Recombinant Rabbit Monoclonal Antibody (ARC1960)

ARF5 Recombinant Monoclonal Antibody for Western Blot, ICC/IF

ADP-ribosylation factor 5 (ARF5) is a member of the human ARF gene family. These genes encode small guanine nucleotide-binding proteins that stimulate the ADP-ribosyltransferase activity of cholera toxin and play a role in vesicular trafficking and as activators of phospholipase D. The gene products include 6 ARF proteins and 11 ARF-like proteins and constitute 1 family of the RAS superfamily. The ARF proteins are categorized as class I (ARF1, ARF2, and ARF3), class II (ARF4 and ARF5) and class III (ARF6). The members of each class share a common gene organization. The ARF5 gene spans approximately 3.2kb of genomic DNA and contains six exons and five introns.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ARF5
Applications ELISA, Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone 0U0D2
Concentration 0.55 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene ARF5
Gene Accession No. P84083, P84084, P84085
Gene Alias ADP ribosylation factor 5; ADP-ribosylation factor 5; ARF5; EFL2; EPLG3; LERK3
Gene Symbols ARF5
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 81-180 of human ARF5 (P84085).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11844, 381, 79117
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.