Learn More
ARF6/ADP Ribosylation Factor 6 Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A305238AT

Description
The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Rat, D. melanogaster, X. laevis, Chicken and Pig
ARF6 (ADP-ribosylation factor 6) is a broadly expressed, differentially regulated GTPase that is present in cytoplasm and on both cell-surface and intracellular membranes and whose functions may include tissue-specific effects on vesicular trafficking during cellular differentiation.Specifications
ARF6/ADP Ribosylation Factor 6 | |
Polyclonal | |
TBS with 0.1% BSA and 0.09% sodium azide | |
P62330, P62331 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
11845, 382 | |
4° C | |
Liquid |
Western Blot | |
Unconjugated | |
ARF6 | |
ARF6 | |
Between 125 and 175 | |
20 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.