Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ARF6 Recombinant Rabbit Monoclonal Antibody (9E7T9)
GREENER_CHOICE

Catalog No. PIMA535119
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA535119 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA535119 Supplier Invitrogen™ Supplier No. MA535119
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Immunogen sequence: HYYTGTQGLI FVVDCADRDR IDEARQELHR IINDREMRDA IILIFANKQD LPDAMKPHEI QEKLGLTRIR DRNWYVQPSC ATSGDGLYEG LTWLTSNYKS.

ARF6 (ADP-ribosylation factor 6) is a broadly expressed, differentially regulated GTPase that is present in cytoplasm and on both cell-surface and intracellular membranes and whose functions may include tissue-specific effects on vesicular trafficking during cellular differentiation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ARF6
Applications ELISA, Western Blot
Classification Recombinant Monoclonal
Clone 9E7T9
Concentration 0.8 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene ARF6
Gene Accession No. P62330, P62331, P62332
Gene Alias ADP ribosylation factor 6; ADP-ribosylation factor 6; AI788669; ARF6; AW496366; DKFZp564M0264
Gene Symbols ARF6
Host Species Rabbit
Immunogen Synthetic Peptide.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11845, 382, 79121
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.